Learn More
Selleck Chemical LLC Lincomycin Hydrochloride Monohydrate S5295-25mg
SDP
Catalog No. 503108229
Encompass_Preferred
Lincomycin hydrochloride monohydrate is a narrow-spectrum antibiotic used in the treatment of staphylococcal streptococcal and Bacteroides fragilis infections
Catalog No. 50-310-8229 Supplier Selleck Chemical LLC Supplier No. S529525MG
Lincomycin hydrochloride monohydrate is a narrow-spectrum antibiotic used in the treatment of staphylococcal streptococcal and Bacteroides fragilis infections