Learn More
Medchemexpress LLC HY-B1358 500mg Medchemexpress, Lincomycin (hydrochloride monohydrate) CAS:7179-49-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1358500MG
Encompass_Preferred
Medchemexpress, HY-B1358 500mg Lincomycin (hydrochloride monohydrate) CAS:7179-49-9 Lincomycin hydrochloride monohydrate is a narrow-spectrum antibiotic, has similar effects to erythromycin, which has a good effect on gram-positive coccus, mainly used to inhibit the synthesis of bacterial cell protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2698