Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lincomycin hydrochloride | 859-18-7 | 99.3% | 1 G
SDP

Catalog No. 5000408926
Encompass_Preferred

Lincomycin hydrochloride is the salt form of the antibiotic lincomycin. It inhibits protein synthesis in gram-positive and, to a lesser extent, gram-negative bacteria by binding to the 50S subunits of bacterial ribosomes and acting on aminoacyl-tRNA binding and the transpeptidation reaction.

  • Inhibits protein synthesis in bacteria
  • Binds to 50S subunits of bacterial ribosomes
  • Active against gram-positive and some gram-negative bacteria
  • Soluble in H2O at 100 mg/mL
  • Transported at room temperature
  • Store at 4°C in sealed storage, away from moisture
  • Can be stored at -80°C for 6 months or -20°C for 1 month when dissolved in solvent

Catalog No. 50-004-08926 Supplier Medchemexpress LLC Supplier No. HYB0417A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lincomycin hydrochloride is the salt form of the antibiotic lincomycin. It inhibits protein synthesis in gram-positive and, to a lesser extent, gram-negative bacteria by binding to the 50S subunits of bacterial ribosomes and acting on aminoacyl-tRNA binding and the transpeptidation reaction.

  • Inhibits protein synthesis in bacteria
  • Binds to 50S subunits of bacterial ribosomes
  • Active against gram-positive and some gram-negative bacteria
  • Soluble in H2O at 100 mg/mL
  • Transported at room temperature
  • Store at 4°C in sealed storage, away from moisture
  • Can be stored at -80°C for 6 months or -20°C for 1 month when dissolved in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.