Learn More
Medchemexpress LLC Lincomycin hydrochloride | 859-18-7 | MFCD00083647 | 99.3% | 500 MG
SDP
Catalog No. 5000409085
Encompass_Preferred
Lincomycin hydrochloride is the salt form of the antibiotic lincomycin. It impacts the synthesis of protein, RNA, and DNA in the mycelium of the lincomycin-producing organism *Act. Roseolus*. Lincomycin inhibits protein synthesis in gram-positive and, to a lesser extent, gram-negative bacteria by binding to the 50S subunits of bacterial ribosomes, affecting aminoacyl-tRNA binding and the transpeptidation reaction.
  • Impacts protein, RNA, and DNA synthesis in *Act. Roseolus* mycelium
  • Inhibits protein synthesis in gram-positive and gram-negative bacteria
  • Binds to 50S subunits of bacterial ribosomes
  • Affects aminoacyl-tRNA binding and transpeptidation reaction
Catalog No. 50-004-09085 Supplier Medchemexpress LLC Supplier No. HYB0417A500MG
Lincomycin hydrochloride is the salt form of the antibiotic lincomycin. It impacts the synthesis of protein, RNA, and DNA in the mycelium of the lincomycin-producing organism *Act. Roseolus*. Lincomycin inhibits protein synthesis in gram-positive and, to a lesser extent, gram-negative bacteria by binding to the 50S subunits of bacterial ribosomes, affecting aminoacyl-tRNA binding and the transpeptidation reaction.
  • Impacts protein, RNA, and DNA synthesis in *Act. Roseolus* mycelium
  • Inhibits protein synthesis in gram-positive and gram-negative bacteria
  • Binds to 50S subunits of bacterial ribosomes
  • Affects aminoacyl-tRNA binding and transpeptidation reaction