Learn More
Description
Specifications
Specifications
| Type | Lincomycin Hydrochloride |
| Chemical Name or Material | Lincomycin Hydrochloride |
| Physical Form | Powder |
| CAS | 859-18-7 |
| Color | White |
| Quantity | 1 g |
| Assay Percent Range | ≥82% |
| Molecular Formula | C18H34N2O6S·HCl |
| Synonym | Lincocin hydrochloride, Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-2-pyrrolidinecarboxamido)-1-thio-D-erythro-α-D-galactooctopyranoside hydrochloride, U-10149A |
| Molecular Weight (g/mol) | 442.996 |
| Show More |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.