Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lincomycin, hydrochloride, ≥82%, MP Biomedicals™

Catalog No. ICN15894805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15894805 5 g
ICN15894801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15894805 Supplier MP Biomedicals, Inc Supplier No. 0215894805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lincomycin, Hydrochloride

Lincomycin hydrochloride is used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding.
TRUSTED_SUSTAINABILITY

Specifications

Type Lincomycin Hydrochloride
Chemical Name or Material Lincomycin hydrochloride
Physical Form Powder
CAS 859-18-7
Color White
Quantity 5 g
Assay Percent Range ≥82%
Molecular Formula C18H34N2O6S·HCl
Synonym Lincocin hydrochloride, Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-2-pyrrolidinecarboxamido)-1-thio-D-erythro-α-D-galactooctopyranoside hydrochloride, U-10149A
Molecular Weight (g/mol) 442.996
Percent Purity ≥82%
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.