Learn More
Electron Microscopy Sciences Lyophilization Lined Aluminum Seals Natural 20 mm O.D., 20 mm O.D.
SDP
Catalog No. 501898958
Encompass_Preferred
Lined lyophilization seals are ideal for filling or retrieving contents with syringe and as closures for many instrumentation applications, center disc tear-out, seal lined with PTFE - Faced natural red rubber, fit mouth OD 20mm.
Catalog No. 50-189-8958 Supplier Electron Microscopy Sciences Supplier No. 6033020N
Lined lyophilization seals are ideal for filling or retrieving contents with syringe and as closures for many instrumentation applications, center disc tear-out, seal lined with PTFE - Faced natural red rubber, fit mouth OD 20mm.