Learn More
Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves
Description
Use Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves with Lindberg/Blue M Tube Furnaces.
Specifications
Specifications
| For Use With (Equipment) | STF55666C Tube Furnace |
| Type | Tube Furnace Adapter, 4 in. |
| Diameter (Metric) Process Tube | 101.6 mm |