Learn More
Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves
Catalog No. p-4529204
Change view
Click to view available options
Type:
Tube Furnace Adapter, 3 in.
Tube Furnace Adapter, 4 in.
For Use With (Equipment):
HTF55322, HTF55342, HTF55347 Furnace
STF55666C Tube Furnace
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
01-257-107 Tube Furnace Adapter, 3 in. HTF55322, HTF55342, HTF55347 Furnace
01-257-101 Tube Furnace Adapter, 4 in. STF55666C Tube Furnace
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-257-107 Supplier Thermo Scientific™ Supplier No. 59525
Only null left
Add to Cart
Add to Cart
Use Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves with Lindberg/Blue M Tube Furnaces.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) HTF55322, HTF55342, HTF55347 Furnace
Type Tube Furnace Adapter, 3 in.
Diameter (Metric) Process Tube 76.2 mm