Learn More
Bioworld Lindemann Orchid Medium, 50 L
Catalog No. 50254640
Encompass_Preferred
Lindemann Orchid MediumContains the Macronutrients, Micronutrients, Sucrose, and Vitamins as described by Lindemann et al. (1970)Cytokines and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 22.71 g/LSoluble in water
Catalog No. 50-254-640 Supplier Bioworld Supplier No. 306300433
Lindemann Orchid MediumContains the Macronutrients, Micronutrients, Sucrose, and Vitamins as described by Lindemann et al. (1970)Cytokines and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 22.71 g/LSoluble in water