Learn More
Grainger Tablecraft Products Company 86 Mesh Strainer, Dia. 6 1/4 in
Catalog No. NC1390266
Encompass_Preferred
The Tablecraft 86 Mesh Strainer is a 6.25 inch diameter stainless steel strainer with an 86 mesh count. It has a depth of 1.5 inches and features a rolled edge rim for durability. The fine mesh allows for straining liquids or sifting dry ingredients. Made from 18/8 stainless steel, it is durable, rust-resistant, and easy to clean. With its fine mesh, this strainer is ideal for straining stocks, broths, sauces, purees, dusting powders, and more in commercial kitchens.
Catalog No. NC1390266 Supplier Grainger Supplier No. 6DVU9
The Tablecraft 86 Mesh Strainer is a 6.25 inch diameter stainless steel strainer with an 86 mesh count. It has a depth of 1.5 inches and features a rolled edge rim for durability. The fine mesh allows for straining liquids or sifting dry ingredients. Made from 18/8 stainless steel, it is durable, rust-resistant, and easy to clean. With its fine mesh, this strainer is ideal for straining stocks, broths, sauces, purees, dusting powders, and more in commercial kitchens.