Learn More
Cayman Chemical Linderalactone, Purity: ≥98%, CAS: 728-61-0, 10 mg
SDP
Catalog No. 502909978
Encompass_Preferred
A sesquiterpene lactone with anticancer and antioxidant activities; an inhibitor of α-glucosidase (IC50 = 12.72 µM); inhibits the proliferation of A549 lung cancer cells (IC50 = 15 µM); also induces apoptosis and cell cycle arrest at the G2/M phase and reduces the levels of unphosphorylated and phosphorylated JAK1, JAK2, STAT1, and STAT2 in A549 cells in a concentration-dependent manner; reduces hydrogen peroxide-induced increases in cell death in HepG2 cells (EC50 = 98 µM)
Catalog No. 50-290-9978 Supplier Cayman Chemical Supplier No. 3564110 MG
A sesquiterpene lactone with anticancer and antioxidant activities; an inhibitor of α-glucosidase (IC50 = 12.72 µM); inhibits the proliferation of A549 lung cancer cells (IC50 = 15 µM); also induces apoptosis and cell cycle arrest at the G2/M phase and reduces the levels of unphosphorylated and phosphorylated JAK1, JAK2, STAT1, and STAT2 in A549 cells in a concentration-dependent manner; reduces hydrogen peroxide-induced increases in cell death in HepG2 cells (EC50 = 98 µM)