Learn More
Medchemexpress LLC HY-N0781 5mg Medchemexpress, Linderalactone CAS:728-61-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07815MG
Encompass_Preferred
Medchemexpress, HY-N0781 5mg Linderalactone CAS:728-61-0 Linderalactone is an important sesquiterpene lactone isolated from Radix linderae. Linderalactone inhibits cancer growth by modulating the expression of apoptosis-related proteins and inhibition of JAK/STAT signalling pathway. Linderalactone also inhibits the proliferation of the lung cancer A-549 cells with an IC50 of 15 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0308