Learn More
Medchemexpress LLC HY-N0688 5mg Medchemexpress, Linderane CAS:13476-25-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06885MG
Encompass_Preferred
Medchemexpress, HY-N0688 5mg Linderane CAS:13476-25-0 Linderane, isolated from the root of Lindera strychnifolia, is an irreversible inhibitor cytochrome P450 2C9 (CYP2C9). Linderane has the potential to relieve pain and cramp. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2810