Learn More
BD Cell Analysis 3P LINEAGE COCKTAIL 1 LIN 1
Supplier:  BD Cell Analysis 3P 340546
Encompass
The Lineage Cocktail 1 (lin 1) contains antibody clones that, in combination, stain lymphocytes, monocytes, eosinophils, and neutrophils. It includes CD3, CD14, CD16, CD19, CD20, and CD56. Peripheral blood dendritic cells and basophils can be distinguished from other leucocytes by their lack of staining with lin 1
Catalog No. 50-620-212