Learn More
Genelink Linear Acrylamide Solution (Linear polyacrylamide, LPA; 5mg/mL); 1 mL
SDP
Supplier:  Genelink 40511301
Encompass_Preferred
Effective DNA and RNA precipitation with the use of monovalent salts and ethanol or isopropanol is dependent on its concentration. DNA and specifically RNA precipitation at a concentration of less than 50 ng/mL is usually not quantitative and the pellet is not clearly visible leading to unreliable recovery. Addition of linear acrylamide as a carrier/coprecipitant aids in quantitative recovery and specifically clear visibility pf the pellet. Linear acrylamide does not interfere with spectrophotometric readings, electrophoresis, and most molecular biology enzymatic applications including PCR. Linear acrylamide is prepared synthetically and is guaranteed free of all nucleic acids, DNase, RNase and proteases.
Catalog No. NC1781917