Learn More
Grainger Warner Linear B-Track K2X Linear Actuator, 230V AC, 3,000 lbf, 8 in. Stroke
Catalog No. 502785361
Encompass_Preferred
The B-Track K2X Linear Actuator is a robust permanent magnet motor-driven unit designed for demanding applications requiring an alternative to hydraulic or air cylinders. This device features an efficient in-line ball screw system and is engineered for reliability in environments where hydraulic power sources are unavailable.
  • Features a high load capacity of 3,000 lbf
  • Equipped with a 230V AC permanent magnet motor with 50/60 Hz frequency
  • Includes an integrated clutch and a capacitor for operational control
  • Utilizes a bronze nut and ball screw drive system for precise movement
  • Constructed with a powder coated aluminum housing and heavy wall design
  • Incorporates automatic thermal protection and a bi-directional wrap spring brake
  • Features a 50% duty cycle with a 0.9 in./sec. speed at full load
Catalog No. 50-278-5361 Supplier Grainger Supplier No. 18G782
The B-Track K2X Linear Actuator is a robust permanent magnet motor-driven unit designed for demanding applications requiring an alternative to hydraulic or air cylinders. This device features an efficient in-line ball screw system and is engineered for reliability in environments where hydraulic power sources are unavailable.
  • Features a high load capacity of 3,000 lbf
  • Equipped with a 230V AC permanent magnet motor with 50/60 Hz frequency
  • Includes an integrated clutch and a capacitor for operational control
  • Utilizes a bronze nut and ball screw drive system for precise movement
  • Constructed with a powder coated aluminum housing and heavy wall design
  • Incorporates automatic thermal protection and a bi-directional wrap spring brake
  • Features a 50% duty cycle with a 0.9 in./sec. speed at full load