Learn More
Takara Bio LINEAR PUROMYCIN MARKER 2 UG
SDP
Catalog No. NC9135878
Encompass
Linear Puromycin Marker, Size: 2microgram, it is a short ds DNA fragment consisting of the puromycin resistance gene regulated by the SV40 promoter and ending with the SV40 polyadenylation signal. Preparation and Storage: At -20degreeC
Catalog No. NC9135878 Supplier Takara Bio Supplier No. 631626
Linear Puromycin Marker, Size: 2microgram, it is a short ds DNA fragment consisting of the puromycin resistance gene regulated by the SV40 promoter and ending with the SV40 polyadenylation signal. Preparation and Storage: At -20degreeC