Learn More
World Precision Instrument Linear Scale Micrometer Ruler 1mm/0.01mm
SDP
Catalog No. 502521441
Encompass
  • 1X3" slide with 1mm linear target etched into the glass down to 10μm marks
  • Linear 1mm/100/0.01mm
  • Marks at 50 and 100μm
  • Suitable for upright microscopes at 40X, 100X and 400X
Catalog No. 50-252-1441 Supplier World Precision Instrument Supplier No. 500828
  • 1X3" slide with 1mm linear target etched into the glass down to 10μm marks
  • Linear 1mm/100/0.01mm
  • Marks at 50 and 100μm
  • Suitable for upright microscopes at 40X, 100X and 400X