Learn More
World Precision Instrument Linear Stimulus Isolator-SYS-A395R
SDP
Catalog No. 502538500
Encompass
Model A395 generates a user-defined output current of wave shape; DC, AC, pulse and combinations. Battery operated, pho­to­elec­tri­cal­ly-isolated from the input voltage drive, the instrument regen­er­ates out­put currents which are linearly proportional to the analog voltage wave­forms pro­vid­ed by your D/A con­vert­er or sig­nal generator.
Catalog No. 50-253-8500 Supplier World Precision Instrument Supplier No. SYSA395R
Model A395 generates a user-defined output current of wave shape; DC, AC, pulse and combinations. Battery operated, pho­to­elec­tri­cal­ly-isolated from the input voltage drive, the instrument regen­er­ates out­put currents which are linearly proportional to the analog voltage wave­forms pro­vid­ed by your D/A con­vert­er or sig­nal generator.