Learn More
Electron Microscopy Sciences EMS Linear (X&Y) Stage Micrometer, Model IAM-2, T/L, Calibrated
SDP
Catalog No. 5034578
Encompass_Preferred
A dual axis linear micrometer that can accurately calibrate both X and Y directions without rotation, for optical magnification, and camera aspect ratio using single standard. In addition, it can be used to calibrate and check linear stage motion or related auto measuring systems. Scales are in both English and Metric measurements. Calibrated.
Catalog No. 50-345-78 Supplier Electron Microscopy Sciences Supplier No. 602162PGC
A dual axis linear micrometer that can accurately calibrate both X and Y directions without rotation, for optical magnification, and camera aspect ratio using single standard. In addition, it can be used to calibrate and check linear stage motion or related auto measuring systems. Scales are in both English and Metric measurements. Calibrated.