Learn More
AUDIT™ MicroControls™ Linearity FD Bilirubin Beckman AU

Description
- Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
- FREE and easy to use Auditor™ QC 'instant reports' data reduction software or AUDIT™ Faxback.
- AUDIT™ Ship provides unrivaled, personalized standing order autoship service.
Specifications
Specifications
| Certifications/Compliance | CLIA-88 |
| Content And Storage | 2°C to 8°C |
| Detects | Direct Bilirubin and Total Bilirubin |
| Manufacturing Origin | US - Aalto Scientific |
| Product Type | Verification Panel |
| Fill Volume | 1 mL |
| Formulation | Freeze Dried |
| For Use With (Application) | Intended to simulate human patient samples for purpose of determining linearity, calibration verification, and verification of reportable range for included analytes |
| For Use With (Equipment) | Beckman AU Analyzers |
| Packaging Type | Kit/Box |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.