Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aalto Scientific Linearity FD Immunoassay for Ortho Vitros
SDP

Catalog No. 22010140
Change view
Click to view available options
Quantity:
10 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-010-140 10 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-010-140 Supplier Aalto Scientific Supplier No. K961M10
Add to Cart
Add to Cart

The Audit MicroControls Linearity FD Immunoassay for Ortho Vitros is designed to simulate human patient samples for linearity studies, calibration verification, and reportable range verification across multiple immunoassay analytes.

  • Intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: AFP, B-HCG, Cortisol, Estradiol, Ferritin, Folate, FSH, Progesterone, Prolactin, Testosterone, TSH, Total PSA, Vitamin B12, LH

Specifications

Detectable Analytes alpha-Fetoprotein (AFP), Cortisol, Estradiol, Ferritin, Folate, FSH, Progesterone, Prolactin, Testosterone, TSH, Total PSA, Vitamin B12, LH
Sample Type Bovine Serum Albumin
Type Linearity FD Immunoassay
Level A to E
Quantity 10 x 5 mL
Form Freeze-dried
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.