Learn More
Aalto Scientific Linearity FD Lipoprotein (a)
SDP
Catalog No. 22022953
Change view
Click to view available options
Quantity:
5 x 1 mL
Catalog No. Quantity
22-022-953 5 x 1 mL
1 options
Catalog No. 22-022-953 Supplier Aalto Scientific Supplier No. K743M5
Add to Cart
Add to Cart
Linearity control material intended to simulate patient samples for linearity, calibration verification, and reportable range verification of lipoprotein (a).
  • The Audit™ MicroControls™ Linearity FD Lipoprotein (a) is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analyte: Lipoprotein (a)
Specifications
Detectable Analytes Lipoprotein (a)
Type Linearity Assay
Quantity 5 x 1 mL
Form Liquid