Learn More
Audit Microcontrol™ Linearity FD Thyroid for Ortho Vitros

Description
- Simulate human patient samples and used in determining linearity, calibration verification, and verify the reportable range for total T3, total T4, free T3, and free T4 analytes.
Specifications
Specifications
| Detectable Analytes | T3 (Free), Total T3, Total T4 |
| Sample Type | Bovine Serum Albumin |
| Type | Linearity, Calibration Verification |
| Level | A to E |
| Quantity | 10 x 2 mL |
| Form | Freeze-dried |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.