Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT™ MicroControls™ Linearity FLQ Special Diabetes for Abbott Systems
SDP

Catalog No. 22158630
Change view
Click to view available options
Quantity:
5 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-630 5 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-630 Supplier AUDIT™ MicroControls™ Supplier No. K956M5

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AUDIT™ MicroControls™, Inc. Linearity FLQ Special Diabetes for Abbott Systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: C-peptide, Insulin.

  • Simulate patient samples for determining linearity, calibration verification, and the verification of reportable range for respective analytes
  • Five distinct levels of C-Peptide and Insulin that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments
  • FREE and easy to use Auditor™ QC “instant reports” data reduction software or AUDIT™ Faxback

Specifications

Detectable Analytes C-peptide, Insulin
Format 5 Vials
For Use With (Equipment) Clinical or IVD Instrumentation
Type Standard Solution Set
For Use With (Application) Instrument Performance Qualification
Product Line FLQ
Quantity 5 x 2 mL
Source Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.