Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT™ MicroControls™ Linearity FLQ TDM for Beckman AU
SDP

Catalog No. 22158626
Change view
Click to view available options
Quantity:
5 x 4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-626 5 x 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-626 Supplier AUDIT™ MicroControls™ Supplier No. K827M5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Linearity FLQ TDM for Beckman AU is assayed quality control material consisting of five levels.

  • Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
  • FREE and easy to use Auditor™ QC 'instant reports' data reduction software or AUDIT™ Faxback.
  • AUDIT™ Ship provides unrivaled, personalized standing order autoship service.

Specifications

Certifications/Compliance CLIA-88
Content And Storage -15°C and then 2°C to 8°C once thawed
Detectable Analytes Acetaminophen, Amikacin, Carbamazepine, Digoxin, Gentamicin, Lidocaine, N-acetylprocainamide (NAPA), Phenobarbital, Phenytoin, Primidone, Procainamide, Quinidine, Salicylate (SALY), Theophylline, Tobramycin, Valproic Acid, Vancomycin
For Use With (Equipment) Beckman AU Analyzers
Type Verification Panel
Disposable Single-use
For Use With (Application) Intended to simulate human patient samples for purpose of determining linearity, calibration verification, and verification of reportable range for included analytes
Level A to E
Packaging Kit
Quantity 5 x 4 mL
Stability Unopened -15°C or colder upon receipt until use, opened 5 days when stored at 2°C to 8°C
Form Frozen Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.