Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT™ MicroControls™ Linearity FLQ Urine/Fluids Chemistry for Beckman AU
SDP

Catalog No. 22158627
Change view
Click to view available options
Quantity:
10 x 3 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-627 10 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-627 Supplier AUDIT™ MicroControls™ Supplier No. K828M5

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Linearity FLQ Urine/Fluids Chemistry for Beckman AU consists of 5 levels.

  • Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments
  • FREE and easy to use Auditor™ QC instant reports data reduction software or AUDIT™ Faxback.

Specifications

Content And Storage Store at -15° and then once thawed, 2-8° for 8 hours
Detectable Analytes Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, Sodium, Phosphorus, Urea Nitrogen, Uric Acid
For Use With (Equipment) Analyzers
Sample Type Human Serum
Type Linearity
For Use With (Application) Intended to Simulate Human Patient Samples for Use in Determining Linearity, Calibration Verification, and The Verification of Reportable Range for The Following Analytes: Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, Sodium, Phosphorous, Urea Nitrogen, Uric Acid.
Quantity 10 x 3 mL
Stability Once thawed, store at 2-8° for 8 hours
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.