Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ HemoScreen Linearity Kit

Catalog No. 23123808
Change view
Click to view available options
Quantity:
5 Levels/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-123-808 5 Levels/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-123-808 Supplier R&D Systems™ Supplier No. HSFA0020100

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Customized HemoScreen Linearity Kit includes 5 levels for calibration verification.

  • For Use With: PixCell™ HemoScreen

Specifications

Certifications/Compliance FDA Licensed Tests
Detectable Analytes Blood, Red Blood Cells, Hemoglobin, Platelets, White Blood Cells
For Use With (Equipment) PixCell™ HemoScreen
Sample Type Whole Blood, Serum, Plasma
Type Linearity Kit
Level 5
Quantity 5 Levels/Box
Stability Stable Through Expiration Date
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.