Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray Linearity Material
SDP

Catalog No. 21377294
Change view
Click to view available options
Sample Type:
Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
21-377-294 Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For in vitro diagnostic use only. Follow lab safety procedures and dispose of materials per regulatory guidelines.
Catalog No. 21-377-294 Supplier Arkray Supplier No. 107253

missing translation for 'validMainframeMsg'

For in vitro diagnostic use only. Follow lab safety procedures and dispose of materials per regulatory guidelines.

ARKRAY Linearity Control Material verifies analyzer performance across key ranges, helping labs ensure accurate results. Compatible with ADAMS systems, ARKRAY Linearity Control Material supports reliable QC and regulatory compliance.

  • Designed for routine linearity verification to support accurate HbA1c analyzer performance.
  • Compatible with ARKRAY ADAMS systems for seamless integration and reliable results.
  • Provides multiple levels to assess instrument response across clinically relevant measurement ranges.
  • Supports laboratory compliance with CLIA, CAP, and internal quality assurance programs.
  • Helps laboratories detect calibration drift, reagent issues, and analytical variation early.
  • Promotes confidence in patient result integrity and clinical decision support.
  • Ready-to-use format simplifies workflow and reduces preparation time.
  • Stabilized composition enhances consistency and long-term storage usability.
  • Ideal for new analyzer verification, routine QC monitoring, or troubleshooting.
  • Backed by ARKRAY expertise in HbA1c systems, ensuring compatibility and dependable performance.

Specifications

Detectable Analytes Hemoglobin
Sample Type Urine
Type Linearity Calibrator
Level 1 Low, 1 Normal, 1 High
Form Liquid

Product performance may vary based on instrument configuration and laboratory procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.