Learn More
VEOLIA WTS LINEARITY STANDARD SET FOR 800
Catalog No. NC9293444
Encompass
Linearity Standards Set - 800/900 Series. Includes 1 each 250 �g C/L, 500 �g C/L, 750 �g C/L as USP Sucrose and 1 reagent water blank.
Catalog No. NC9293444 Supplier VEOLIA WTS Supplier No. STD 3101201
Linearity Standards Set - 800/900 Series. Includes 1 each 250 �g C/L, 500 �g C/L, 750 �g C/L as USP Sucrose and 1 reagent water blank.