Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Checkers Linebacker 1.25 in. Max Diameter ADA Ramps LB125-RMP Checkers

Catalog No. 19088466 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-088-466 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-088-466 Supplier Checkers Supplier No. CPRP1X125BLU
Add to Cart
Add to Cart

General Purpose 1-Channel Linebacker Cable Protectors with 1.25 in. channel size. Stable enough to support both pedestrian traffic as well as carts, forklifts, and heavy trucks. Each pair of ramps is created with easy installation in mind.

  • Patented, modular ADA/DDA Compliant ramps
  • Add ramps and rails when requirements specify
  • One person installation - no tools required

Specifications

Color Blue
Dimensions (L x W x H) 38.5 x 21.75 x 2.25 in.
Material Polyurethane
Product Type Cable Protector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.