Learn More
Lakeside Manufacturing Inc Linen Utility Cart, 4 Shelf: 21" x 33", 500-Lb. Capacity
Catalog No. 502884435
Encompass_Preferred
Linen Carts by Lakeside Manufacturing are compact and easily maneuverable that come in 3 different shelf sizes to meet your unique needs. The Lakeside 443 Linen Utility Cart with (4) shelves measuring 21" x 33" has a weight capacity of 500-Lb. This Lakeside cart's stainless steel construction is easy to clean and sanitize. Its all-welded construction eliminates nuts and bolts for a stronger structure and easier-to-clean surface. This medical utility cart's compact size allows for easy maneuverability. Attached with bumpers on legs and handles to protect walls and furniture. Equipped with 2 fixed and 2 swivel 5” casters that guide cart in a straight line, while making turns easier. Optional nylon cart cover available to protect contents from airborne dust and dirt while still allowing easy accessibility. Modify or custom-build with no minimum orders. Superior lead times. Made in the USA.
Catalog No. 50-288-4435 Supplier Lakeside Manufacturing Inc Supplier No. 443
Linen Carts by Lakeside Manufacturing are compact and easily maneuverable that come in 3 different shelf sizes to meet your unique needs. The Lakeside 443 Linen Utility Cart with (4) shelves measuring 21" x 33" has a weight capacity of 500-Lb. This Lakeside cart's stainless steel construction is easy to clean and sanitize. Its all-welded construction eliminates nuts and bolts for a stronger structure and easier-to-clean surface. This medical utility cart's compact size allows for easy maneuverability. Attached with bumpers on legs and handles to protect walls and furniture. Equipped with 2 fixed and 2 swivel 5” casters that guide cart in a straight line, while making turns easier. Optional nylon cart cover available to protect contents from airborne dust and dirt while still allowing easy accessibility. Modify or custom-build with no minimum orders. Superior lead times. Made in the USA.