Learn More
Trajan Scientific LINER AG 4MM ID QW PKT25
SDP
Catalog No. 502530171
Encompass_Preferred
Trajan GC Inlet Liners, Includes: Split liner; w/quartz wool, Diameter Inner: 4 mm, Diameter Outer: 6.3 mm, Overall Length: 78.5 mm
Catalog No. 50-253-0171 Supplier Trajan Scientific Supplier No. 092220
Trajan GC Inlet Liners, Includes: Split liner; w/quartz wool, Diameter Inner: 4 mm, Diameter Outer: 6.3 mm, Overall Length: 78.5 mm