Learn More
Phenomenex Inc Liner for Shimadzu (3.4mm ID x 95mm L x 5mm OD)
Catalog No. 502412672 Shop All Phenomenex Products
Encompass_Preferred
Liner for Shimadzu (3.4mm ID x 95mm L x 5mm OD) Split/Splitless, Single Taper w/Wool, FocusLiner
Catalog No. 50-241-2672 Supplier Phenomenex Inc Supplier No. U428
Liner for Shimadzu (3.4mm ID x 95mm L x 5mm OD) Split/Splitless, Single Taper w/Wool, FocusLiner