Learn More
Trajan Scientific Liner Inlet Liner ConnecTite Shimadzu 5/PK
SDP
Catalog No. 5000493579
Encompass_Preferred
SGE ConnecTite Shimadzu inlet liners with bottom hole length 95 mm PK5
Catalog No. 50-004-93579 Supplier Trajan Scientific Supplier No. 092331
SGE ConnecTite Shimadzu inlet liners with bottom hole length 95 mm PK5