Learn More
Trajan Scientific Liner Inlet Liner Double Taper Agilent 5/PK
SDP
Catalog No. 5000494890
Encompass_Preferred
SGE double taper inlet liners 4 mm ID length 78 5 mm PK5
Catalog No. 50-004-94890 Supplier Trajan Scientific Supplier No. 092018
SGE double taper inlet liners 4 mm ID length 78 5 mm PK5