Learn More
Trajan Scientific Liner Inlet Liner Focus Shimadzu 5/PK
SDP
Catalog No. 5000494926
Encompass_Preferred
SGE Shimadzu FocusLiners 3 4 mm ID length 99 mm PK5
Catalog No. 50-004-94926 Supplier Trajan Scientific Supplier No. 092065
SGE Shimadzu FocusLiners 3 4 mm ID length 99 mm PK5