Learn More
Trajan Scientific Liner Inlet Liner Focus Taper 5/PK
SDP
Catalog No. 5000494910
Encompass_Preferred
SGE PerkinElmer taper FocusLiners 4 mm ID length 92 mm PK5
Catalog No. 50-004-94910 Supplier Trajan Scientific Supplier No. 092095
SGE PerkinElmer taper FocusLiners 4 mm ID length 92 mm PK5