Learn More
Trajan Scientific Liner Inlet Liner Focus Taper 5/PK
SDP
Catalog No. 5000494895
Encompass_Preferred
SGE Shimadzu taper FocusLiners 3 4 mm ID length 95 mm PK5
Catalog No. 50-004-94895 Supplier Trajan Scientific Supplier No. 092058
SGE Shimadzu taper FocusLiners 3 4 mm ID length 95 mm PK5