Learn More
Trajan Scientific Liner Inlet Liner Gooseneck PerkinElmer 5/PK
SDP
Catalog No. 5000494897
Encompass_Preferred
SGE PerkinElmer gooseneck inlet liners 3 mm ID length 100 mm PK5
Catalog No. 50-004-94897 Supplier Trajan Scientific Supplier No. 092091
SGE PerkinElmer gooseneck inlet liners 3 mm ID length 100 mm PK5