Learn More
Trajan Scientific Liner Inlet Liner Gooseneck Shimadzu 5/PK
SDP
Catalog No. 5000493661
Encompass_Preferred
SGE Shimadzu gooseneck inlet liners 3 4 mm ID length 95 mm PK5
Catalog No. 50-004-93661 Supplier Trajan Scientific Supplier No. 092085
SGE Shimadzu gooseneck inlet liners 3 4 mm ID length 95 mm PK5