Learn More
Trajan Scientific Liner Inlet Liner Gooseneck Shimadzu 5/PK
SDP
Catalog No. 5000493618
Encompass_Preferred
SGE Shimadzu mid-gooseneck inlet liners 3 4 mm ID length 99 mm PK5
Catalog No. 50-004-93618 Supplier Trajan Scientific Supplier No. 092082
SGE Shimadzu mid-gooseneck inlet liners 3 4 mm ID length 99 mm PK5