Learn More
Trajan Scientific Liner Inlet Liner PTV PerkinElmer 5/PK
SDP
Catalog No. 5000494115
Encompass_Preferred
SGE PerkinElmer gooseneck inlet liners PTV 1 mm ID length 88 mm PK5
Catalog No. 50-004-94115 Supplier Trajan Scientific Supplier No. 092097
SGE PerkinElmer gooseneck inlet liners PTV 1 mm ID length 88 mm PK5