Learn More
Trajan Scientific Liner Inlet Liner Shimadzu Medical 5/PK
SDP
Catalog No. 5000493642
Encompass_Preferred
SGE Shimadzu straight inlet liners 0 75 mm ID length 95 mm PK5
Catalog No. 50-004-93642 Supplier Trajan Scientific Supplier No. 092089
SGE Shimadzu straight inlet liners 0 75 mm ID length 95 mm PK5