Learn More
Trajan Scientific Liner Inlet Liner Straight Agilent 25/PK
SDP
Catalog No. 5000494540
Encompass_Preferred
SGE straight inlet liners 4 mm ID length 78 5 mm PK25
Catalog No. 50-004-94540 Supplier Trajan Scientific Supplier No. 092222
SGE straight inlet liners 4 mm ID length 78 5 mm PK25