Learn More
Trajan Scientific Liner Inlet Liner Straight Fison 5/PK
SDP
Catalog No. 5000494929
Encompass_Preferred
SGE Thermo Scientific straight inlet liners 5 mm ID length 105 mm PK5
Catalog No. 50-004-94929 Supplier Trajan Scientific Supplier No. 092150
SGE Thermo Scientific straight inlet liners 5 mm ID length 105 mm PK5