Learn More
Trajan Scientific Liner Inlet Liner Straight Fison 5/PK
SDP
Catalog No. 5000494896
Encompass_Preferred
SGE Thermo Scientific straight inlet liners 3 mm ID length 105 mm PK5
Catalog No. 50-004-94896 Supplier Trajan Scientific Supplier No. 092147
SGE Thermo Scientific straight inlet liners 3 mm ID length 105 mm PK5