Learn More
Trajan Scientific Liner Inlet Liner Straight Fison 5/PK
SDP
Supplier:  Trajan Scientific 092148
Encompass_Preferred
SGE Thermo Scientific straight inlet liners 0 8 mm ID length 105 mm PK5
Catalog No. 50-004-94385