Learn More
Trajan Scientific Liner Inlet Liner Straight PerkinElmer 25/PK
SDP
Catalog No. 5000494095
Encompass_Preferred
SGE PerkinElmer straight inlet liners 4 mm ID length 92 mm PK25
Catalog No. 50-004-94095 Supplier Trajan Scientific Supplier No. 09210025
SGE PerkinElmer straight inlet liners 4 mm ID length 92 mm PK25